Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitro-2-methylimidazole, MP Biomedicals

Catalog No. ICN225229 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225229 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225229 Supplier MP Biomedicals, Inc Supplier No. 0522522910

Chemical Identifiers

CAS 696-23-1
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00005191
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
PubChem CID 12760
IUPAC Name 2-methyl-5-nitro-1H-imidazole
SMILES CC1=NC=C(N1)[N+]([O-])=O

Specifications

Quantity 10 g
Chemical Name or Material 5-Nitro-2-Methyl Imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.