CAS RN 70-18-8
L-Glutathione, reduced, 98+%
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
L-Glutathione, reduced, 97%
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
L-Glutathione, Reduced, 98.0-101.0%, Spectrum™ Chemical
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.32 InChI Key: RWSXRVCMGQZWBV-UHFFFAOYNA-N IUPAC Name: 2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid SMILES: NC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
Glutathione Reduced, Cell Culture Reagent, ≥98%, MP Biomedicals™
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Glutathione Reduced Crystalline MP Biomedicals
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Glutathione, 98%, for analysis, reduced
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.32 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Glutathione reduced form 97.0+%, TCI America™
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 MDL Number: MFCD00065939 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Glutathione Reduced, Fisher BioReagents
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N
Fisher Science Education™ Glutathione
CAS: 70-18-8 Molecular Formula: C10H17N3O6S Molecular Weight (g/mol): 307.321 InChI Key: RWSXRVCMGQZWBV-WDSKDSINSA-N Synonym: glutathione,l-glutathione,glutathion,glutathione-sh,glutinal,isethion,tathion,reduced glutathione,deltathione,neuthion PubChem CID: 124886 ChEBI: CHEBI:16856 IUPAC Name: (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid SMILES: C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N