Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Reduced, Fisher BioReagents

Catalog No. BP252150 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP252150 50 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP252150

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Glutathione is a useful tripeptide involved in many aspects of metabolism, including transport of γ-glutamyl amino acids and reductive cleavage of disulfide bonds.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 182°C
Color White
pH 3
Ignition Residue 0.01% max.
Loss on Drying Report %
Assay Percent Range ≥98 %
Packaging Amber Glass
Quantity 50 g
Merck Index 15, 4511
Specific Rotation -21 to -11° (+ or -)
Formula Weight 307.3
Percent Purity ≥98%
Physical Form Powder or Solid
Chemical Name or Material Glutathione Reduced
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.