accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 700-58-3

IUPAC Name: adamantan-2-one
Synonyms: adamantanone tricyclo(3.3.1.13,7)decanone (9CI) tricyclo(3.3.1.1(sup 3,7))decanone
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: IYKFYARMMIESOX-UHFFFAOYSA-N
SMILES: O=C1C2CC3CC(C2)CC1C3
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (6)
  • (1)
Quantity 
  • (1)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (4)
  • (6)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (6)