accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3NOO

CAS RN 70024-51-0

IUPAC Name: 2-(heptan-3-yl)-5-methyl-5-nitro-1,3-dioxane
Synonyms: 2-(heptan-3-yl)-5-methyl-5-nitro-1,3-dioxane
Molecular Weight (g/mol): 245.32
Molecular Formula: C12H23NO4
InChi Key: FCGHBWUKOGKLKO-UHFFFAOYNA-N
SMILES: CCCCC(CC)C1OCC(C)(CO1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results