accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOHO

CAS RN 701-54-2

IUPAC Name: 4-(aminomethyl)cyclohexane-1-carboxylic acid
Synonyms: tranexamic acid
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: GYDJEQRTZSCIOI-UHFFFAOYSA-N
SMILES: NCC1CCC(CC1)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results