accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOHOOO

CAS RN 70145-54-9

IUPAC Name: sodium (5-hydroxy-4-oxo-4H-pyran-2-yl)methanolate
Synonyms: sodium (5-hydroxy-4-oxopyran-2-yl)methanolate
Molecular Weight (g/mol): 164.09
Molecular Formula: C6H5NaO4
InChi Key: XDGSAVMIBSOBNL-UHFFFAOYSA-N
SMILES: [Na+].OC1=COC(C[O-])=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results