accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70197-13-6

IUPAC Name: trioxorhenium methanide
Synonyms: methylrhenium trioxide
Molecular Weight (g/mol): 249.24
Molecular Formula: CH3O3Re
InChi Key: VZSXFJPZOCRDPW-UHFFFAOYSA-N
SMILES: [CH3-].O=[Re](=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)