Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltrioxorhenium(VII) 97.0+%, TCI America™
SDP

Catalog No. M1296100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1296100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1296100MG Supplier TCI America Supplier No. M1296100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ≥97.0% methyltrioxorhenium(VII) for use as a laboratory reagent.

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re-
Molecular Weight (g/mol) 249.239
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O

Specifications

Melting Point 114°C
Physical Form Crystalline Powder
Quantity 100 mg
Formula Weight 249.24
Percent Purity ≥97.0% (T)
Grade Reagent
Chemical Name or Material Methyltrioxorhenium(VII)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.