accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7021-09-2

IUPAC Name: 2-methoxy-2-phenylacetic acid
Synonyms: methoxy(phenyl)acetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILES: COC(C(O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (4)
Physical Form 
  • (6)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (6)
  • (1)
  • (2)