Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyphenylacetic Acid, MilliporeSigma™

Catalog No. M1597120010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass Bottle Pre Packed
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1597120010 10 g Glass Bottle Pre Packed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1597120010 Supplier MilliporeSigma Supplier No. 1.59712.0010
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7021-09-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
PubChem CID 107202

Specifications

Melting Point 68°C to 72°C
Color White
Packaging Glass Bottle Pre Packed
Quantity 10 g
Formula Weight 166.18
Physical Form Crystalline Powder or Powder
EINECSNumber 230-300-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.