Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-α-Methoxyphenylacetic acid, Solstice

Catalog No. 6002800 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-028-00 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-028-00 Supplier Solstice Supplier No. 6521025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for gravimetric determination of Na,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7021-09-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00064217
InChI Key DIWVBIXQCNRCFE-UHFFFAOYNA-N
PubChem CID 107202
IUPAC Name 2-methoxy-2-phenylacetic acid

Specifications

Packaging Plastic Bottle
Linear Formula C6H5CH(OCH3)COOH
Quantity 25 g
UN Number NONH for all modes of transport
Beilstein 2209197
Synonym methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid
SMILES COC(C1=CC=CC=C1)C(=O)O
Formula Weight 166.17g/mol
Percent Purity ≥99% (T)
Chemical Name or Material (±)-α-Methoxyphenylacetic acid
Hazard Statement P261-P305 + P351 + P338
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.