accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFONHOHN

CAS RN 703-95-7

IUPAC Name: 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
Synonyms: 5-fluoroorotic acid
Molecular Weight (g/mol): 174.09
Molecular Formula: C5H3FN2O4
InChi Key: SEHFUALWMUWDKS-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(=O)NC(=O)N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results