Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroorotic Acid, MP Biomedicals

Catalog No. ICN15421091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15421091 1 g
ICN15421050 50 mg
ICN15421083 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15421091 Supplier MP Biomedicals, Inc Supplier No. 0215421091
Add to Cart
Add to Cart

Chemical Identifiers

CAS 703-95-7
Molecular Formula C5H3FN2O4
Molecular Weight (g/mol) 174.087
MDL Number MFCD00150658
InChI Key SEHFUALWMUWDKS-UHFFFAOYSA-N
Synonym 5-fluoroorotic acid, 5-fluoroorotate, fluoroorotic acid, 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid, orotic acid, 5-fluoro, 5-foa, 5-fluoro orotic acid, 5-fluorouracil-4-carboxylic acid, unii-7ia9ouc93e, 5-fluoroorotic acid van
PubChem CID 69711
ChEBI CHEBI:42535
IUPAC Name 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid
SMILES C1(=C(NC(=O)NC1=O)C(=O)O)F

Specifications

Color Yellow
Assay Percent Range 98.3%
Quantity 1 g
Formula Weight 174.1
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroorotic Acid, Selective agent for yeast molecular genetics and uracil auxotrophs
EINECSNumber 211-876-0
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.