accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70331-82-7

IUPAC Name: sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
Synonyms: sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethanesulfonate
Molecular Weight (g/mol): 251.23
Molecular Formula: C6H14NNaO6S
InChi Key: GUXMFAHOYNRHBI-UHFFFAOYSA-M
SMILES: [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
Physical Form 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results