Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid Sodium Salt Hydrate 99.0+%, TCI America™
SDP

Catalog No. T096225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T096225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T096225G Supplier TCI America Supplier No. T096225G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
Synonym tes sodium salt, sodium 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonate, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid sodium salt, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-, monosodium salt, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-, sodium salt 1:1, tes-sodium salt
PubChem CID 23702152
IUPAC Name sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 251.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid Sodium Salt Hydrate [Good′s buffer comp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.