accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 704-10-9

IUPAC Name: 1-(2,4-dichloro-5-fluorophenyl)ethan-1-one
Synonyms: 1-(2,4-dichloro-5-fluorophenyl)ethanone
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H5Cl2FO
InChi Key: FAKJFAMIABOKBW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)