Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-fluoroacetophenone, 99%, Thermo Scientific™

Catalog No. AC270160250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC270160250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC270160250 Supplier Thermo Scientific Chemicals Supplier No. 270160250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 704-10-9
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD00077499
InChI Key FAKJFAMIABOKBW-UHFFFAOYSA-N
Synonym 2',4'-dichloro-5'-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethanone, 2,4-dichloro-5-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethan-1-one, dcfa, acetophenone, 2',4'-dichloro-5'-fluoro, 6-acetyl-1,3-dichloro-4-fluorobenzene, 1-2,4-dichloro-5-fluoro-phenyl ethanone, ethanone, 1-2,4-dichloro-5-fluorophenyl, 1-acetyl-2,4-dichloro-5-fluorobenzene
PubChem CID 727250
IUPAC Name 1-(2,4-dichloro-5-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC(F)=C(Cl)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 25 g
Formula Weight 207.03
Percent Purity ≥98.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2, 4-Dichloro-5-fluoroacetophenone, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.