Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dichloro-5'-fluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. D238525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D238525G Supplier TCI America Supplier No. D238525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 704-10-9
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD00077499
InChI Key FAKJFAMIABOKBW-UHFFFAOYSA-N
Synonym 2',4'-dichloro-5'-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethanone, 2,4-dichloro-5-fluoroacetophenone, 1-2,4-dichloro-5-fluorophenyl ethan-1-one, dcfa, acetophenone, 2',4'-dichloro-5'-fluoro, 6-acetyl-1,3-dichloro-4-fluorobenzene, 1-2,4-dichloro-5-fluoro-phenyl ethanone, ethanone, 1-2,4-dichloro-5-fluorophenyl, 1-acetyl-2,4-dichloro-5-fluorobenzene
PubChem CID 727250
IUPAC Name 1-(2,4-dichloro-5-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC(F)=C(Cl)C=C1Cl

Specifications

Melting Point 35°C
Color White
Boiling Point 115°C
Quantity 25 g
Formula Weight 207.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dichloro-5′-fluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.