accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7057-57-0

IUPAC Name: zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
Synonyms: zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
Molecular Weight (g/mol): 447.06
Molecular Formula: C18H15Cl3N2OZn
InChi Key: YTEJSAFVYHDCSN-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C=C3
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (3)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)