Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Dimethylamino-2,3-Benzophenoxazine Hemi (Zinc Chloride) Salt, MP Biomedicals™

Catalog No. ICN15778805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15778805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15778805 Supplier MP Biomedicals, Inc Supplier No. 0215778805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7057-57-0
Molecular Formula C18H15ClN2O
Molecular Weight (g/mol) 310.781
InChI Key HWYNRVXFYFQSID-UHFFFAOYSA-M
Synonym basic blue 6, meldola's blue, phenylene blue, unii-cp2st8ed1o, c.i. basic blue 6, cp2st8ed1o, 9-dimethylamino benzo a phenoxazin-7-ium chloride, benzo a phenoxazin-7-ium, 9-dimethylamino-, chloride, 8-dimethylamino-2,3-benzophenoxazine hemi zinc chloride salt, n,n-dimethyl-10-oxa-5-azatetraphen-8-iminium chloride
PubChem CID 101929
IUPAC Name benzo[a]phenoxazin-9-ylidene(dimethyl)azanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=CC4=CC=CC=C43)OC2=C1)C.[Cl-]

Specifications

Quantity 5 g
Formula Weight 378.9
Physical Form Solid
Chemical Name or Material 8-Dimethylamino-2, 3-Benzophenoxazine Hemi (Zinc Chloride) Salt
Recommended Storage Store at Room Temperature (15deg to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.