Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meldola's Blue, Thermo Scientific™

Catalog No. AC334090050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC334090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC334090050 Supplier Thermo Scientific Chemicals Supplier No. 334090050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7057-57-0
Molecular Formula C18H15Cl3N2OZn
Molecular Weight (g/mol) 447.06
InChI Key YTEJSAFVYHDCSN-UHFFFAOYSA-K
Synonym Basic Blue 6, C.I. 51175
PubChem CID 101929
IUPAC Name zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
SMILES [Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C=C3

Specifications

Formula Weight 310.78
Quantity 5 g
Chemical Name or Material Meldola's Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.