accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70788-28-2

IUPAC Name: N-(diaminophosphoryl)-4-fluorobenzamide
Synonyms: flurofamide flurofamidum flurfamide 氟法胺 flurofamida
Molecular Weight (g/mol): 217.14
Molecular Formula: C7H9FN3O2P
InChi Key: QWZFVMCWPLMLTL-UHFFFAOYSA-N
SMILES: NP(N)(=O)NC(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results