accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHONHBrBrH3C

CAS RN 708275-58-5

IUPAC Name: 1-(2,4-dibromophenyl)-3-(2,2-dimethyl-4-phenyl-1,3-dioxan-5-yl)urea
Synonyms: 1-(2,4-dibromophenyl)-3-(2,2-dimethyl-4-phenyl-1,3-dioxan-5-yl)urea
Molecular Weight (g/mol): 484.19
Molecular Formula: C19H20Br2N2O3
InChi Key: RBKIJGLHFFQHBE-UHFFFAOYNA-N
SMILES: CC1(C)OCC(NC(=O)NC2=C(Br)C=C(Br)C=C2)C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)