accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 70931-28-1

IUPAC Name: 4-[(4-fluorophenyl)methyl]piperazine-1,4-diium
Synonyms: 4-[(4-fluorophenyl)methyl]piperazine-1,4-diium
Molecular Weight (g/mol): 196.27
Molecular Formula: C11H17FN2
InChi Key: OOSZCNKVJAVHJI-UHFFFAOYSA-P
SMILES: FC1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results