Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorobenzyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH6199314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6199314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6199314 Supplier Thermo Scientific Chemicals Supplier No. H6199314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70931-28-1
Molecular Formula C11H17FN2
Molecular Weight (g/mol) 196.27
MDL Number MFCD01075234
InChI Key OOSZCNKVJAVHJI-UHFFFAOYSA-P
Synonym 1-4-fluorobenzyl piperazine, 1-4-fluorophenyl methyl piperazine, 1-4-fluorobenzyl-piperazine, piperazine,1-4-fluorophenyl methyl, 1-4-fluoro-benzyl-piperazine, 4-fluorobenzylpiperazine, piperazine, 1-4-fluorophenyl methyl, 4-fluorophenyl methyl piperazine, 4-fluorobenzyl-piperazine, acmc-1bl7k
PubChem CID 796563
IUPAC Name 1-[(4-fluorophenyl)methyl]piperazine
SMILES FC1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1

Specifications

Melting Point 63°C to 67°C
Quantity 25 g
UN Number UN3259
Formula Weight 194.25
Percent Purity 97%
Chemical Name or Material 1-(4-Fluorobenzyl)piperazine
Hazard Category H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.