accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71-67-0

IUPAC Name: disodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
Synonyms: bromsulphthalein|bromsulfalein|bromosulphthalein|bromosulfalein|sodium sulfobromophthalein|sulphobromophthalein sodium|tetrabromophenolsulfophthalein|bromosulfophthalein sodium|sodium bromosulfalein|sodium sulphobromophthalein
Molecular Weight (g/mol): 837.99
Molecular Formula: C20H8Br4Na2O10S2
InChi Key: GHAFORRTMVIXHS-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O
Physical Form 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)