Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfobromophthalein Sodium Salt, MP Biomedicals™

Catalog No. MP215672601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215672601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215672601 Supplier MP Biomedicals, Inc Supplier No. 0215672601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a substrate for multidrug resistance associated protein 2 (Mrp2). Sulfobromophthalein Sodium Salt, MP Biomedicals is a dye with a high affinity for organic anion transporting polypeptides (OATPs). BSP is used in diagnosis of hepatic disorders and

BSP is transported into hepatocytes by OATPs and, after conjugation to glutathione, is excreted into bile by Mrp2.3 It was found to inhibit the aldo-keto reductase ARK1C20.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-67-0
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD00150017
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady
PubChem CID 102371197
ChEBI CHEBI:63827
IUPAC Name disodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O

Specifications

Quantity 1 g
Solubility Information Soluble in water (20mg/mL, clear, colorless solution). Insoluble in ethanol or acetone.
Formula Weight 838
Physical Form Powder
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.