accessibility menu, dialog, popup

UserName

Viewing profile as user:
SeNOO

CAS RN 71098-88-9

IUPAC Name: 2-(phenylselanyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(phenylselanyl)isoindole-1,3-dione
Molecular Weight (g/mol): 302.2
Molecular Formula: C14H9NO2Se
InChi Key: TYZFQSLJTWPSDS-UHFFFAOYSA-N
SMILES: O=C1N([Se]C2=CC=CC=C2)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results