accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONSOOONa

CAS RN 71119-22-7

IUPAC Name: sodium 3-(morpholin-4-yl)propane-1-sulfonate
Synonyms: sodium 3-(morpholin-4-yl)propane-1-sulfonate
Molecular Weight (g/mol): 231.24
Molecular Formula: C7H14NNaO4S
InChi Key: MWEMXEWFLIDTSJ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCCN1CCOCC1
Physical Form 
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (4)
  • (3)
  • (2)
Grade 
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)