Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS Sodium Salt Crystalline MP Biomedicals

Catalog No. ICN19067091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19067091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19067091 Supplier MP Biomedicals, Inc Supplier No. 0219067091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zwitterionic 'Good' buffer suitable for biological applications at a pH range closer to 7.4.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-[N-Morpholino]propanesulfonic acid
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 231.2
Grade Biochemical
Chemical Name or Material MOPS Sodium Salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.