accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOONO

CAS RN 71119-23-8

IUPAC Name: sodium 2-(morpholin-4-yl)ethane-1-sulfonate
Synonyms: mes-na sodium mes mes sodium salt
Molecular Weight (g/mol): 217.21
Molecular Formula: C6H12NNaO4S
InChi Key: IRHWMYKYLWNHTL-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCN1CCOCC1
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (6)
Grade 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)