Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MES Sodium Salt, MP Biomedicals™

Catalog No. ICN15245480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15245480 100 g
ICN15245410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15245480 Supplier MP Biomedicals, Inc Supplier No. 0215245480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71119-23-8
Molecular Formula C6H12NNaO4S
Molecular Weight (g/mol) 217.215
MDL Number MFCD00065473
InChI Key IRHWMYKYLWNHTL-UHFFFAOYSA-M
Synonym mes sodium salt, sodium 2-morpholinoethanesulfonate, 2-n-morpholino ethanesulfonic acid sodium salt, 4-morpholineethanesulfonic acid, sodium salt, sodium 4-morpholin-1-ylethylsulphonate, sodium 2-4-morpholinyl ethanesulfonate, mes-na, 4-morpholineethanesulfonic acid sodium salt, sodium 4-morpholineethanesulfonate, 4-morpholineethanesulfonic acid, sodium salt 1:1
PubChem CID 23673676
ChEBI CHEBI:62955
IUPAC Name sodium;2-morpholin-4-ylethanesulfonate
SMILES C1COCCN1CCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Assay Percent Range 1.003
Quantity 100 g
Formula Weight 217.2
Grade Not Available
Physical Form Powder
Chemical Name or Material MES
EINECSNumber 275-203-2
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.