accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71160-24-2

IUPAC Name: (6E,8E,10E,14E)-5,12-dihydroxyicosa-6,8,10,14-tetraenoic acid
Synonyms: 5,12-DiHETE
Molecular Weight (g/mol): 336.47
Molecular Formula: C20H32O4
InChi Key: VNYSSYRCGWBHLG-XUOUMLBJNA-N
SMILES: CCCCC\C=C\CC(O)\C=C\C=C\C=C\C(O)CCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results