accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNN(E/Z)(E/Z)

CAS RN 71274-65-2

IUPAC Name: N'-[(3-methylphenyl)amino]-N-(phenylimino)benzenecarboximidamide
Synonyms: N'-[(3-methylphenyl)amino]-N-(phenylimino)benzenecarboximidamide
Molecular Weight (g/mol): 314.39
Molecular Formula: C20H18N4
InChi Key: TXSUIVPRHHQNTM-UHFFFAOYSA-N
SMILES: CC1=CC(NN=C(N=NC2=CC=CC=C2)C2=CC=CC=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results