accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7144-65-2

IUPAC Name: 2-({[1,1'-biphenyl]-2-yloxy}methyl)oxirane
Synonyms: 2-({[1,1'-biphenyl]-2-yloxy}methyl)oxirane
Molecular Weight (g/mol): 226.28
Molecular Formula: C15H14O2
InChi Key: DNVXWIINBUTFEP-UHFFFAOYNA-N
SMILES: C(OC1=CC=CC=C1C1=CC=CC=C1)C1CO1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)