accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOOHOOHOHOH

CAS RN 71484-85-0

IUPAC Name: 3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylic acid
Synonyms: 3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylic acid
Molecular Weight (g/mol): 315.23
Molecular Formula: C12H13NO9
InChi Key: QSUILVWOWLUOEU-UHFFFAOYNA-N
SMILES: OC1C(O)C(OC2=CC=C(C=C2)[N+]([O-])=O)OC(C1O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results