accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHOHNOOHO

CAS RN 7164-43-4

IUPAC Name: 5-amino-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
Synonyms: 5-amino-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylic acid
Molecular Weight (g/mol): 171.11
Molecular Formula: C5H5N3O4
InChi Key: HWCXJKLFOSBVLH-UHFFFAOYSA-N
SMILES: NC1=C(NC(=O)NC1=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results