Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoorotic Acid 97.0+%, TCI America™
SDP

Catalog No. A094725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A094725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A094725G Supplier TCI America Supplier No. A094725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7164-43-4
Molecular Formula C5H5N3O4
Molecular Weight (g/mol) 171.11
MDL Number MFCD00010563
InChI Key HWCXJKLFOSBVLH-UHFFFAOYSA-N
Synonym 5-Aminouracil-6-carboxylic Acid
PubChem CID 73018
IUPAC Name 5-amino-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
SMILES NC1=C(NC(=O)NC1=O)C(O)=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 171.11
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoorotic Acid
RTECSNumber RM3190000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.