accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNNFFFNHHNOSOOCH3

CAS RN 717906-29-1

IUPAC Name: N-methyl-N-{2-[({2-[(2-oxo-2,3-dihydro-1H-indol-5-yl)amino]-5-(trifluoromethyl)pyrimidin-4-yl}amino)methyl]phenyl}methanesulfonamide
Synonyms: N-methyl-N-{2-[({2-[(2-oxo-1,3-dihydroindol-5-yl)amino]-5-(trifluoromethyl)pyrimidin-4-yl}amino)methyl]phenyl}methanesulfonamide
Molecular Weight (g/mol): 506.5
Molecular Formula: C22H21F3N6O3S
InChi Key: POJZIZBONPAWIV-UHFFFAOYSA-N
SMILES: CN(C1=CC=CC=C1CNC1=NC(NC2=CC3=C(NC(=O)C3)C=C2)=NC=C1C(F)(F)F)S(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)