CAS RN 72093-04-0
CAS RN 72093-04-0
IUPAC Name:
3-chloro-4-methylpyridine
Synonyms:
3-chloro-4-methylpyridine
Molecular Weight (g/mol):
127.57
Molecular Formula:
C6H6ClN
InChi Key:
VLNXNJMHLZFECV-UHFFFAOYSA-N
SMILES:
CC1=C(Cl)C=NC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3-Chloro-4-methylpyridine, 95%, Thermo Scientific™
CAS: 72093-04-0 Molecular Formula: C6H6ClN Molecular Weight (g/mol): 127.57 MDL Number: MFCD04114245 InChI Key: VLNXNJMHLZFECV-UHFFFAOYSA-N Synonym: 3-chloro-4-picoline,pyridine, 3-chloro-4-methyl,3-chloro-4-methyl-pyridine,pubchem14323,acmc-1bl3c,3-chloro-4-methylpyridine?,ksc494c8t,3-chloro-4-methylpyridine PubChem CID: 2762925 IUPAC Name: 3-chloro-4-methylpyridine SMILES: CC1=C(Cl)C=NC=C1