accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3OHNH3C

CAS RN 721-50-6

IUPAC Name: N-(2-methylphenyl)-2-(propylamino)propanamide
Synonyms: prilocaine propitocaine prilocaine preparation citanest plain prilocaine (substance) prilocaina 丙胺卡因 prilocainum prilocaine base citanest octapressin
Molecular Weight (g/mol): 220.32
Molecular Formula: C13H20N2O
InChi Key: MVFGUOIZUNYYSO-UHFFFAOYNA-N
SMILES: CCCNC(C)C(=O)NC1=CC=CC=C1C
Percent Purity 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (6)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (6)