accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(R)(R)Br(R)HONOHNO

CAS RN 72218-68-9

IUPAC Name: 1-[(2R,3R,4R,5R)-3-bromo-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[(2R,3R,4R,5R)-3-bromo-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 307.1
Molecular Formula: C9H11BrN2O5
InChi Key: FEUDNSHXOOLCEY-XVFCMESISA-N
SMILES: OC[C@H]1O[C@H]([C@H](Br)[C@@H]1O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results