accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NH2

CAS RN 72233-91-1

IUPAC Name: 1-methyl-1-(3-methylphenyl)hydrazine
Synonyms: 1-methyl-1-(3-methylphenyl)hydrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: ABANUKHECQXXRH-UHFFFAOYSA-N
SMILES: CN(N)C1=CC=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results