accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NFCl

CAS RN 72235-56-4

IUPAC Name: 1-(3-chloro-4-fluorophenyl)methanamine
Synonyms: 1-(3-chloro-4-fluorophenyl)methanamine
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: LQAUXDMGRBWDIU-UHFFFAOYSA-N
SMILES: NCC1=CC=C(F)C(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results