Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluorobenzylamine 98.0+%, TCI America™
SDP

Catalog No. C27025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27025G 5 g
C27021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27025G Supplier TCI America Supplier No. C27025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72235-56-4
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD00042507
InChI Key LQAUXDMGRBWDIU-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzylamine, 3-chloro-4-fluorophenyl methanamine, 3-chloro-4-fluorobenzyl amine, 1-3-chloro-4-fluorophenyl methanamine, benzenemethanamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenemethanamine, 3-chloro-4-fluorophenyl methylamine, pubchem4375, acmc-209omc, 3-chloro4-fluorobenzylamine
PubChem CID 144539
IUPAC Name (3-chloro-4-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)Cl)F

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
UN Number 2735
Formula Weight 159.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.