accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNHSOOH2N

CAS RN 723-46-6

IUPAC Name: 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzene-1-sulfonamide
Synonyms: sulfamethoxazole sulfametoxazol sulfamethoxazole - chemical sulphamethoxazole sulpha-methoxizole sulfamethoxazol sulfamethoxazole (product) sulfisomezole sulfamethoxazole preparation sulphamethylisoxazole
Molecular Weight (g/mol): 253.28
Molecular Formula: C10H11N3O3S
InChi Key: JLKIGFTWXXRPMT-UHFFFAOYSA-N
SMILES: CC1=CC(NS(=O)(=O)C2=CC=C(N)C=C2)=NO1
Physical Form 
  • (5)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (4)
Type 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)