Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamethoxazole, 50mg/mL in DMSO, sterile-filtered, Thermo Scientific™

Catalog No. AAJ67346AD
Change view
Click to view available options
Quantity:
25 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67346AD 50 mL
AAJ67346AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67346AD Supplier Thermo Scientific Chemicals Supplier No. J67346AD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723-46-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
MDL Number MFCD00010546
InChI Key JLKIGFTWXXRPMT-UHFFFAOYSA-N
PubChem CID 5329
ChEBI CHEBI:9332
IUPAC Name 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
SMILES CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Type Sulfamethoxazole
Physical Form Liquid
Packaging Plastic Bottle
Quantity 50 mL
Chemical Name or Material Sulfamethoxazole
Hazard Category H335
EINECSNumber 211-963-3
RTECSNumber WP0700000
TSCA Yes
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.