CAS RN 723331-20-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ZJ 43
Glutamate carboxypeptidase II and III (NAALADase, NAAG peptidase) inhibitor