Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZJ 43

Catalog No. 26751R Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-751-R 1 mg
26-751-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-751-R Supplier Tocris Bioscience™ Supplier No. 2675/1
This item is not returnable. View return policy

Glutamate carboxypeptidase II and III (NAALADase, NAAG peptidase) inhibitor

Potent inhibitor of glutamate carboxypeptidase II and III (GCP II and III/NAAG peptidase/NAALADase) (Ki values are 0.8 and 23nM respectively) that inhibits the hydrolysis of NAAG (IC50 = 2.4nM) . Does not directly interact with NMDA or metabotropic glutamate receptors. Reduces neuronal degeneration in a rat model of? traumatic brain injury (TBI) and reduces locomotor activity in the PCP-model of schizophrenia.

Specifications

Chemical Name or Material N-[[[(1S)-1-Carboxy-3-methylbutyl]amino]carbonyl]-L-glutamic acid
CAS 723331-20-2
Quantity 1 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C12H20N2O7
Formula Weight Observed MW: 304.3

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.